siRNA Details
Gene Symbol: Pa2g4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004206.1 | NP_001004206.1 | 3 | 416 | ||
GenBank mRNA | BC079095.1 | - | 3 | 416 | 7124036 | |
FQ233061.1 | - | 3 | 424 |

More Information
Other Species Targeted

Gene Ontology Categories:
