siRNA Details
Gene Symbol: Ormdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105940.1 | NP_001099410.1 | 3 | 489 | ||
XM_006240753.2 | XP_006240815.1 | 3 | 469 | |||
XM_006240754.2 | XP_006240816.1 | 3 | 389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
