siRNA Details
Gene Symbol: Olr883 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001358.1 | NP_001001358.1 | 2 | 645 |
Gene Symbol: LOC100362926 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729779.2 | XP_002729825.2 | 2 | 669 |
Gene Symbol: Olr910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001359.1 | NP_001001359.1 | 2 | 645 |
