siRNA Details
Gene Symbol: Neurod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105942.1 | NP_001099412.1 | 2 | 995 | ||
XM_006240818.2 | XP_006240880.1 | 4 | 1385 | |||
XM_006240819.2 | XP_006240881.1 | 4 | 1518 | |||
XM_006240820.2 | XP_006240882.1 | 3 | 1297 | |||
XM_006240822.2 | XP_006240884.1 | 4 | 1261 | |||
XM_006240823.2 | XP_006240885.1 | 2 | 1031 |
