siRNA Details
Gene Symbol: Olr1002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001371.1 | NP_001001371.1 | 1 | 631 |
Gene Symbol: Olr956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001381.1 | NP_001001381.1 | 1 | 631 |
