siRNA Details
Gene Symbol: Cc2d1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013869.1 | NP_001013891.1 | 16 | 1861 | ||
XM_006255233.2 | XP_006255295.1 | 17 | 2053 | |||
XM_006255234.2 | XP_006255296.1 | 17 | 2050 | |||
XM_008772364.1 | XP_008770586.1 | 16 | 1919 | |||
GenBank mRNA | BC081948.1 | - | 16 | 1861 | 7122187 |

More Information
Other Species Targeted

Gene Ontology Categories:
