siRNA Details
Gene Symbol: LOC288913 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198728.1 | NP_942023.1 | 3 | 495 | ||
GenBank mRNA | X62277.1 | - | 3 | 495 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
