siRNA Details
Gene Symbol: Trmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013870.1 | NP_001013892.1 | 2, 3 | 287 | ||
XM_006255236.2 | XP_006255298.1 | 3, 4 | 2517 | |||
XM_008772365.1 | XP_008770587.1 | 3, 4 | 2593 | |||
XM_008772366.1 | XP_008770588.1 | 2, 3 | 2378 | |||
GenBank mRNA | BC085126.1 | - | 2, 3 | 287 | 7132178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
