siRNA Details
Gene Symbol: Wdr83os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255237.1 | XP_006255299.1 | 2, 3 | 337 | ||
GenBank mRNA | BC166543.1 | - | 2, 3 | 213 | 7300989 | |
FQ225008.1 | - | 2, 3 | 342 |

More Information
Other Species Targeted

