siRNA Details
Gene Symbol: Wdr83os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255237.1 | XP_006255299.1 | 3 | 409 | ||
GenBank mRNA | BC166543.1 | - | 3 | 285 | 7300989 | |
FQ225008.1 | - | 3 | 414 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
