siRNA Details
Gene Symbol: Serpinb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105948.1 | NP_001099418.1 | 7 | 1018 | ||
XM_008769428.1 | XP_008767650.1 | 7 | 1280 | |||
XM_008769429.1 | XP_008767651.1 | 8 | 1852 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
