siRNA Details
Gene Symbol: Kiss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181692.1 | NP_859043.1 | 2 | 314 | ||
XM_008769443.1 | XP_008767665.1 | 3 | 767 | |||
GenBank mRNA | AY196983.1 | - | 2 | 314 | ||
JX139030.1 | - | 3 | 725 | |||
JX139031.1 | - | 3 | 521 | |||
JX139032.1 | - | 3 | 446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
