siRNA Details
Gene Symbol: Adipor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207587.1 | NP_997470.1 | 7 | 1067 | ||
XM_006249852.2 | XP_006249914.1 | 7 | 1103 | |||
XM_008769543.1 | XP_008767765.1 | 7 | 1404 | |||
XM_008769544.1 | XP_008767766.1 | 7 | 1766 | |||
GenBank mRNA | BC061838.1 | - | 7 | 1067 | 5599938 | |
DQ148391.1 | - | 6 | 855 |

More Information
Other Species Targeted

Gene Ontology Categories:
