siRNA Details
Gene Symbol: Edem3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191671.1 | NP_001178600.1 | 9 | 1005 | ||
XM_006249985.2 | XP_006250047.1 | 9 | 1439 | |||
XM_006249986.2 | XP_006250048.1 | 9 | 1439 | |||
XM_006249987.2 | XP_006250049.1 | 9 | 1439 | |||
XM_008769612.1 | XP_008767834.1 | 9 | 1439 |
