siRNA Details
Gene Symbol: Ivns1abp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047085.1 | NP_001040550.1 | 7 | 1030 | ||
XM_006249988.2 | XP_006250050.1 | 12 | 1643 | |||
GenBank mRNA | AY553870.2 | - | 7 | 1030 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001039512, NM_054102, AK145071, AK146645, AK144619, and others.
Gene Ontology Categories:
