siRNA Details
Gene Symbol: Ivns1abp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047085.1 | NP_001040550.1 | 7 | 982 | ||
XM_006249988.2 | XP_006250050.1 | 12 | 1595 | |||
GenBank mRNA | AY553870.2 | - | 7 | 982 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
