siRNA Details
Gene Symbol: Acbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011906.1 | NP_001011906.1 | 2 | 739 | ||
XM_006250031.2 | XP_006250093.1 | 2 | 795 | |||
XM_006250032.2 | XP_006250094.1 | 2 | 803 | |||
XM_006250033.2 | XP_006250095.1 | 2 | 802 | |||
XM_006250035.2 | XP_006250097.1 | 2 | 796 | |||
XM_006250036.2 | XP_006250098.1 | 2 | 796 | |||
GenBank mRNA | BC086598.1 | - | 2 | 739 | 7313105 |
