siRNA Details
Gene Symbol: Acbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011906.1 | NP_001011906.1 | 2 | 735 | ||
XM_006250031.2 | XP_006250093.1 | 2 | 791 | |||
XM_006250032.2 | XP_006250094.1 | 2 | 799 | |||
XM_006250033.2 | XP_006250095.1 | 2 | 798 | |||
XM_006250035.2 | XP_006250097.1 | 2 | 792 | |||
XM_006250036.2 | XP_006250098.1 | 2 | 792 | |||
GenBank mRNA | BC086598.1 | - | 2 | 735 | 7313105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
