siRNA Details
Gene Symbol: Cacybp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004208.1 | NP_001004208.1 | 4 | 539 | ||
GenBank mRNA | BC079007.1 | - | 4 | 539 | 7113129 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009786, AK089043, AK212050, AK215280, U97327, and others.
Gene Ontology Categories:
