siRNA Details
Gene Symbol: Cacybp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004208.1 | NP_001004208.1 | 2, 3 | 397 | ||
GenBank mRNA | BC079007.1 | - | 2, 3 | 397 | 7113129 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009786, AK089043, AK145013, U97327, AK013924, and others.
Gene Ontology Categories:
