siRNA Details
Gene Symbol: Cenpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033061.2 | NP_001028233.2 | 4 | 776 | ||
NM_001244761.1 | NP_001231690.1 | 3 | 690 | |||
XM_006250099.2 | XP_006250161.1 | 4 | 828 | |||
XM_006250100.2 | XP_006250162.1 | 3 | 741 | |||
GenBank mRNA | BC079467.1 | - | 4 | 776 | 7122576 | |
BC089893.1 | - | 3 | 690 | |||
BC103642.1 | - | 4 | 769 | 7300924 |

More Information
Other Species Targeted

