siRNA Details
Gene Symbol: Mettl11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_222824.7 | XP_222824.5 | 3 | 831 | ||
GenBank mRNA | FQ213916.1 | - | 2 | 576 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
