siRNA Details
Gene Symbol: Mpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077643.1 | NP_001071111.1 | 3, 4 | 243 | ||
XM_006250126.2 | XP_006250188.1 | 3, 4 | 1038 | |||
GenBank mRNA | FQ215073.1 | - | 3, 4 | 384 | ||
FQ217960.1 | - | 3, 4 | 312 | |||
FQ222522.1 | - | 3, 4 | 378 | |||
FQ224252.1 | - | 3, 4 | 275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
