siRNA Details
Gene Symbol: Creg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105966.1 | NP_001099436.1 | 3 | 589 | ||
XM_008769625.1 | XP_008767847.1 | 3 | 713 | |||
GenBank mRNA | BC166446.1 | - | 3 | 603 | 8364240 | |
FQ226272.1 | - | 3 | 575 | |||
FQ227495.1 | - | 3 | 593 | |||
FQ230214.1 | - | 3 | 622 | |||
FQ230343.1 | - | 3 | 587 | |||
FQ231443.1 | - | 3 | 576 | |||
FQ232342.1 | - | 3 | 593 | |||
FQ234634.1 | - | 3 | 622 | |||
FQ234923.1 | - | 3 | 586 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
