siRNA Details
Gene Symbol: Ppox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105968.1 | NP_001099438.1 | 11 | 1432 | ||
XM_006250240.2 | XP_006250302.1 | 10 | 2333 | |||
XM_006250241.1 | XP_006250303.1 | 11 | 1497 | |||
XM_006250243.1 | XP_006250305.1 | 10 | 1140 |
