siRNA Details
Gene Symbol: Nit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082580.1 | NP_001076049.1 | 2 | 170 | ||
NM_182668.2 | NP_872609.2 | 2 | 138 | |||
GenBank mRNA | BC087146.1 | - | 2 | 119 | 7129801 | |
CB576182.1 | - | 2 | 233 | |||
CO558072.1 | - | 2 | 176 | |||
FQ214471.1 | - | 168 | ||||
FQ227281.1 | - | 167 |
