siRNA Details
Gene Symbol: TRPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135955.1 | NP_001129427.1 | 2 | 501 | ||
NM_001135956.1 | NP_001129428.1 | 2 | 501 | |||
NM_001135957.1 | NP_001129429.1 | 2 | 501 | |||
NM_001135958.1 | NP_001129430.1 | 2 | 501 | |||
NM_003306.1 | NP_003297.1 | 2 | 501 | |||
NM_016179.2 | NP_057263.1 | 2 | 501 | |||
XM_017020724.1 | XP_016876213.1 | 2 | 1073 | |||
GenBank mRNA | AF063822.1 | - | 2 | 501 | ||
AF063823.1 | - | 2 | 501 | |||
AF063824.1 | - | 2 | 501 | |||
AF063825.1 | - | 2 | 501 | |||
AF175406.1 | - | 2 | 497 | |||
AF421358.1 | - | 1 | 266 | |||
AF421359.1 | - | 1 | 266 | |||
AF421360.1 | - | 1 | 266 | |||
AF421361.1 | - | 1 | 266 | |||
AF421362.1 | - | 1 | 266 | |||
AK312524.1 | - | 2 | 471 | |||
BC104725.1 | - | 2 | 422 | 40008478 | ||
BC105306.1 | - | 2 | 422 | |||
BC105641.1 | - | 2 | 422 | 40008481 | ||
BC105917.1 | - | 2 | 422 | 40008477 | ||
BX648312.1 | - | 2 | 1125 | |||
DB258184.1 | - | 2 | 491 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
