siRNA Details
Gene Symbol: TRPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135958.1 | NP_001129430.1 | 10 | 2124 | ||
NM_003306.1 | NP_003297.1 | 11 | 2658 | |||
NM_016179.2 | NP_057263.1 | 11 | 2643 | |||
XM_011535206.1 | XP_011533508.1 | 8 | 1278 | |||
XM_017020723.1 | XP_016876212.1 | 8 | 1278 | |||
XM_017020724.1 | XP_016876213.1 | 11 | 3204 | |||
GenBank mRNA | AF063822.1 | - | 11 | 2643 | ||
AF175406.1 | - | 11 | 2639 | |||
AF421358.1 | - | 10 | 2408 | |||
AF421360.1 | - | 10 | 2423 | |||
AF421361.1 | - | 9 | 2071 | |||
AF421362.1 | - | 9 | 1889 | |||
AK312524.1 | - | 11 | 2613 | |||
BC104725.1 | - | 11 | 2564 | 40008478 | ||
BC105641.1 | - | 11 | 2564 | 40008481 | ||
BC105917.1 | - | 11 | 2564 | 40008477 | ||
BX648312.1 | - | 11 | 3267 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
