siRNA Details
Gene Symbol: Olr1601 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000087.1 | NP_001000087.1 | 1 | 223 |
Gene Symbol: LOC103693685 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769806.1 | XP_008768028.1 | 223 |

More Information
Other Species Targeted

Gene Ontology Categories:
