siRNA Details
Gene Symbol: Grem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105974.1 | NP_001099444.1 | 2 | 523 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011825, AK161827, BC079905, AB011030, AK142008.
Gene Ontology Categories:
