siRNA Details
Gene Symbol: Tfb2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008293.1 | NP_001008294.1 | 4 | 728 | ||
XM_006250361.2 | XP_006250423.1 | 4 | 770 | |||
XM_008769817.1 | XP_008768039.1 | 4 | 772 | |||
GenBank mRNA | BC085870.1 | - | 4 | 728 | 7191198 |

More Information
Other Species Targeted

Gene Ontology Categories:
