siRNA Details
Gene Symbol: Tfb2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008293.1 | NP_001008294.1 | 1 | 377 | ||
XM_006250361.2 | XP_006250423.1 | 1 | 419 | |||
XM_008769817.1 | XP_008768039.1 | 1 | 421 | |||
GenBank mRNA | BC085870.1 | - | 1 | 377 | 7191198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
