siRNA Details
Gene Symbol: Rps6kc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105984.1 | NP_001099454.1 | 10 | 1520 | ||
XM_006250459.2 | XP_006250521.1 | 11 | 1699 | |||
XM_006250460.2 | XP_006250522.1 | 10 | 1606 | |||
XM_006250461.2 | XP_006250523.1 | 11 | 1838 | |||
XM_008769829.1 | XP_008768051.1 | 11 | 1699 | |||
XM_008769830.1 | XP_008768052.1 | 10 | 1339 | |||
XM_008769831.1 | XP_008768053.1 | 10 | 1476 | |||
XM_008769832.1 | XP_008768054.1 | 4 | 4050 | |||
XM_008769833.1 | XP_008768055.1 | 4 | 735 | |||
XM_008769834.1 | XP_008768056.1 | 5 | 669 | |||
GenBank mRNA | BC064438.1 | - | 11 | 1770 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
