siRNA Details
Gene Symbol: Rps6kc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105984.1 | NP_001099454.1 | 9, 10 | 1200 | ||
XM_006250459.2 | XP_006250521.1 | 10, 11 | 1379 | |||
XM_006250460.2 | XP_006250522.1 | 9, 10 | 1286 | |||
XM_006250461.2 | XP_006250523.1 | 10, 11 | 1518 | |||
XM_008769829.1 | XP_008768051.1 | 10, 11 | 1379 | |||
XM_008769830.1 | XP_008768052.1 | 9, 10 | 1019 | |||
XM_008769831.1 | XP_008768053.1 | 9, 10 | 1156 | |||
XM_008769832.1 | XP_008768054.1 | 3, 4 | 3730 | |||
XM_008769833.1 | XP_008768055.1 | 3, 4 | 415 | |||
XM_008769834.1 | XP_008768056.1 | 4, 5 | 349 | |||
GenBank mRNA | BC064438.1 | - | 10, 11 | 1450 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
