siRNA Details
Gene Symbol: G0s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009632.1 | NP_001009632.1 | 2 | 166 | ||
GenBank mRNA | BC088248.1 | - | 2 | 166 | 7369006 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
