siRNA Details
Gene Symbol: Cr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105989.2 | NP_001099459.2 | 7 | 1267 | ||
XM_006250476.2 | XP_006250538.1 | 11 | 1882 | |||
XM_006250477.2 | XP_006250539.1 | 7 | 1336 | |||
XM_008769847.1 | XP_008768069.1 | 14 | 2467 | |||
XM_008769848.1 | XP_008768070.1 | 7 | 1753 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
