siRNA Details
Gene Symbol: Ephx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105994.1 | NP_001099464.1 | 6 | 749 | ||
XM_006250541.1 | XP_006250603.1 | 6 | 802 | |||
XM_008769927.1 | XP_008768149.1 | 6 | 1661 | |||
GenBank mRNA | FQ213206.1 | - | 6 | 675 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
