siRNA Details
Gene Symbol: Naa11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024742.1 | NP_001019913.1 | 1 | 452 | ||
XM_006250697.2 | XP_006250759.1 | 1 | 473 | |||
GenBank mRNA | BC078912.1 | - | 1 | 425 | ||
BC097350.1 | - | 1 | 452 | 7462336 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
