siRNA Details
Gene Symbol: Mrpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105997.2 | NP_001099467.1 | 3 | 315 | ||
XM_008770009.1 | XP_008768231.1 | 4 | 682 | |||
GenBank mRNA | BC168772.1 | - | 3 | 315 | 6920150 | |
FQ217255.1 | - | 3 | 317 |
Gene Symbol: LOC100359687 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750509.3 | XP_003750557.1 | 3 | 309 |

More Information
Other Species Targeted

Gene Ontology Categories:
