siRNA Details
Gene Symbol: Ccni | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105998.1 | NP_001099468.1 | 8 | 1502 | ||
XM_006250715.2 | XP_006250777.1 | 7 | 1602 | |||
GenBank mRNA | FQ223715.1 | - | 7 | 1595 | ||
FQ225999.1 | - | 1 | 187 |
