siRNA Details
Gene Symbol: Ccni | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105998.1 | NP_001099468.1 | 8 | 1498 | ||
XM_006250715.2 | XP_006250777.1 | 7 | 1598 | |||
GenBank mRNA | FQ223715.1 | - | 7 | 1591 | ||
FQ225999.1 | - | 1 | 183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
