siRNA Details
Gene Symbol: Prr27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250795.2 | XP_006250857.1 | 5 | 726 | ||
XM_006250796.2 | XP_006250858.1 | 5 | 726 | |||
XM_006250797.2 | XP_006250859.1 | 4 | 702 |

More Information
Other Species Targeted

