siRNA Details
Gene Symbol: Igfbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013048.1 | NP_001013066.1 | 2, 3 | 609 | ||
GenBank mRNA | BC086582.1 | - | 2, 3 | 609 | 7307584 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
