siRNA Details
Gene Symbol: Ociad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013874.1 | NP_001013896.1 | 7 | 690 | ||
XM_006250894.2 | XP_006250956.1 | 8 | 736 | |||
GenBank mRNA | BC083718.1 | - | 7 | 690 | 7190773 | |
FQ212940.1 | - | 7 | 655 | |||
FQ213030.1 | - | 7 | 699 | |||
FQ213629.1 | - | 7 | 727 |

More Information
Other Species Targeted

