siRNA Details
Gene Symbol: Ube2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106006.1 | NP_001099476.1 | 1, 2 | 281 | ||
GenBank mRNA | FQ226880.1 | - | 1, 2 | 273 | ||
FQ233155.1 | - | 1, 2 | 229 | |||
FQ234076.1 | - | 1, 2 | 259 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_016786, AK048194, AK036068, BC002013, AK166647, and others.
Gene Ontology Categories:
