siRNA Details
Gene Symbol: Tbc1d19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106008.2 | NP_001099478.1 | 9 | 712 | ||
GenBank mRNA | BC166575.1 | - | 9 | 712 | 7321142 |

More Information
Other Species Targeted

Gene Ontology Categories:
