siRNA Details
Gene Symbol: Gba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251045.2 | XP_006251107.1 | 2, 3 | 381 | ||
XM_006251046.2 | XP_006251108.1 | 3, 4 | 484 | |||
XM_006251047.2 | XP_006251109.1 | 2, 3 | 382 | |||
XM_008770205.1 | XP_008768427.1 | 2, 3 | 382 | |||
GenBank mRNA | BC168903.1 | - | 1, 2 | 160 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
