siRNA Details
Gene Symbol: Tada2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170455.1 | NP_001163926.1 | 2 | 341 | ||
XM_008770258.1 | XP_008768480.1 | 2 | 1056 | |||
GenBank mRNA | BC169109.1 | - | 2 | 341 | 7315785 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
