siRNA Details
Gene Symbol: Rhbdd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013875.1 | NP_001013897.1 | 8 | 1540 | ||
XM_006251204.2 | XP_006251266.1 | 9 | 1637 | |||
XM_006251205.2 | XP_006251267.1 | 8 | 1427 | |||
XM_006251206.2 | XP_006251268.1 | 9 | 1470 | |||
XM_006251207.2 | XP_006251269.1 | 8 | 1428 | |||
XM_006251208.2 | XP_006251270.1 | 9 | 1577 | |||
XM_006251209.2 | XP_006251271.1 | 8 | 1620 | |||
XM_006251210.2 | XP_006251272.1 | 7 | 1416 | |||
GenBank mRNA | BC081912.1 | - | 8 | 1540 | 7112018 |
