siRNA Details
Gene Symbol: Upp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030025.1 | NP_001025196.1 | 6 | 563 | ||
GenBank mRNA | BC099752.1 | - | 6 | 563 | 5623209 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Upp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030025.1 | NP_001025196.1 | 6 | 563 | ||
GenBank mRNA | BC099752.1 | - | 6 | 563 | 5623209 |